Product Details

SNP ID
rs116520407
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:15171573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCACTGTTAAGCTATTTTAAATG[A/C]AATGATATTTATTTATCTTCTGAAA
Phenotype
MIM: 613574
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TTC39B PubMed Links

Gene Details

Gene
TTC39B
Gene Name
tetratricopeptide repeat domain 39B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168339.1 2827 UTR 3 NP_001161811.1
NM_001168340.1 2827 UTR 3 NP_001161812.1
NM_001168341.1 2827 UTR 3 NP_001161813.1
NM_001168342.1 2827 UTR 3 NP_001161814.1
NM_152574.2 2827 UTR 3 NP_689787.2
XM_011517732.2 2827 UTR 3 XP_011516034.1
XM_017014310.1 2827 UTR 3 XP_016869799.1
XM_017014311.1 2827 UTR 3 XP_016869800.1
XM_017014312.1 2827 UTR 3 XP_016869801.1

View Full Product Details