Product Details
- SNP ID
-
rs117579945
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:15564102 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGGAAAACATCATGAATTTGAATA[C/T]TTCAAGTAATACTGGTGATACCCAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC171
PubMed Links
Gene Details
- Gene
- CCDC171
- Gene Name
- coiled-coil domain containing 171
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173550.2 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
NP_775821.2 |
XM_005251397.2 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
XP_005251454.1 |
XM_005251398.3 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
XP_005251455.1 |
XM_005251399.4 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
XP_005251456.1 |
XM_011517788.2 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
XP_011516090.1 |
XM_011517791.2 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
XP_011516093.1 |
XM_011517796.2 |
329 |
Intron |
|
|
XP_011516098.1 |
XM_017014431.1 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
XP_016869920.1 |
XM_017014432.1 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
XP_016869921.1 |
XM_017014433.1 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
XP_016869922.1 |
XM_017014434.1 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
XP_016869923.1 |
XM_017014435.1 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
XP_016869924.1 |
XM_017014436.1 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
XP_016869925.1 |
XM_017014437.1 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
XP_016869926.1 |
XM_017014438.1 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
XP_016869927.1 |
XM_017014439.1 |
329 |
UTR 5 |
|
|
XP_016869928.1 |
XM_017014440.1 |
329 |
Intron |
|
|
XP_016869929.1 |
XM_017014441.1 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
XP_016869930.1 |
XM_017014442.1 |
329 |
Missense Mutation |
ACT,ATT |
T5I |
XP_016869931.1 |
XM_017014443.1 |
329 |
Intron |
|
|
XP_016869932.1 |
XM_017014444.1 |
329 |
Intron |
|
|
XP_016869933.1 |
View Full Product Details