Product Details

SNP ID
rs112053518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:20346477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGATGTTTCCAGGTAACTCTGT[A/G]GTTTACGGACTGTGGTTTTGTCCAG
Phenotype
MIM: 159558
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MLLT3 PubMed Links

Gene Details

Gene
MLLT3
Gene Name
MLLT3, super elongation complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286691.1 1727 Missense Mutation CCA,CTA P555L NP_001273620.1
NM_004529.3 1727 Missense Mutation CCA,CTA P558L NP_004520.2
XM_017014726.1 1727 Missense Mutation CCA,CTA P510L XP_016870215.1
XM_017014727.1 1727 Missense Mutation CCA,CTA P507L XP_016870216.1

View Full Product Details