Product Details

SNP ID
rs116748628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:26904445 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATTGCTAGGATGTTCACACACAT[C/T]TGTGATCTCTCTAGAAAGTATAGCA
Phenotype
MIM: 603873
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLAA PubMed Links

Gene Details

Gene
PLAA
Gene Name
phospholipase A2 activating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031689.2 3813 UTR 3 NP_001026859.1
NM_001321546.1 3813 UTR 3 NP_001308475.1
XM_011518072.2 3813 Intron XP_011516374.1
XM_017015319.1 3813 Intron XP_016870808.1

View Full Product Details