Product Details

SNP ID
rs113363499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:27524592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTATGAAGAGGGACATCAAGAAG[A/G]CCTTCTATGAAATGTCCCTACAGGC
Phenotype
MIM: 615326
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFNK PubMed Links

Gene Details

Gene
IFNK
Gene Name
interferon kappa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020124.2 279 Missense Mutation ACC,GCC T86A NP_064509.2
Gene
MOB3B
Gene Name
MOB kinase activator 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024761.4 279 Intron NP_079037.3

View Full Product Details