Product Details
- SNP ID
-
rs111261393
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:32558803 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATACACAAAACAGGGACTTAAACC[A/G]AAAGTGAGAAGGAAAGAACATTTTA
- Phenotype
-
MIM: 603322
MIM: 609507
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NDUFB6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142936680] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NDUFB6
- Gene Name
- NADH:ubiquinone oxidoreductase subunit B6
- Gene
- TOPORS
- Gene Name
- TOP1 binding arginine/serine rich protein
There are no transcripts associated with this gene.
- Gene
- TOPORS-AS1
- Gene Name
- TOPORS antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details