Product Details

SNP ID
rs115524004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33461790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCAGAATTACAGGAAGCAGAGA[A/G]GCTACCAGAAAGTGGGTGTAGACCA
Phenotype
MIM: 611532
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6851 PubMed Links

Gene Details

Gene
MIR6851
Gene Name
microRNA 6851
There are no transcripts associated with this gene.

Gene
NOL6
Gene Name
nucleolar protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022917.4 4416 UTR 3 NP_075068.2
NM_139235.3 4416 UTR 3 NP_631981.2
XM_017015044.1 4416 UTR 3 XP_016870533.1

View Full Product Details