Product Details

SNP ID
rs115948946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33463292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAGGAGGATCATAGCCCAGCAC[A/G]GGCATCAGGGATGAGGGCCCCGGCT
Phenotype
MIM: 611532
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6851 PubMed Links

Gene Details

Gene
MIR6851
Gene Name
microRNA 6851
There are no transcripts associated with this gene.

Gene
NOL6
Gene Name
nucleolar protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022917.4 3245 Silent Mutation CCC,CCT P1048P NP_075068.2
NM_139235.3 3245 Intron NP_631981.2
XM_017015044.1 3245 Silent Mutation CCC,CCT P441P XP_016870533.1

View Full Product Details