Product Details

SNP ID
rs115285410
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34092339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCACCATGCCACATCTTAGCAT[A/G]AATAGAATTTTAGTTCAGGCAGGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAF12 PubMed Links

Gene Details

Gene
DCAF12
Gene Name
DDB1 and CUL4 associated factor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015397.3 Intron NP_056212.1
XM_011517843.1 Intron XP_011516145.1

View Full Product Details