Product Details

SNP ID
rs117028434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34379110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGATAGTCGGGATAGGTCTCAGAC[A/G]CAAACTCAGGATGGATAACATAGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf24 PubMed Links

Gene Details

Gene
C9orf24
Gene Name
chromosome 9 open reading frame 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252195.1 887 Missense Mutation GCG,GTG A154V NP_001239124.1
NM_032596.3 887 UTR 3 NP_115985.2
NM_147168.1 887 UTR 3 NP_671697.1
NM_147169.2 887 Missense Mutation GCG,GTG A101V NP_671698.1
XM_005251616.4 887 Missense Mutation GCG,GTG A289V XP_005251673.1
XM_005251618.4 887 Missense Mutation GCG,GTG A154V XP_005251675.1
XM_011518055.2 887 Missense Mutation GCG,GTG A154V XP_011516357.1
Gene
KIAA1161
Gene Name
KIAA1161
There are no transcripts associated with this gene.

View Full Product Details