Product Details

SNP ID
rs113042654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35674194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACCCAGAGAGGAGGATCCACCC[A/G]GAGAGGAGGATCTACCTGGAGAGGA
Phenotype
MIM: 603179 MIM: 190990
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF39 PubMed Links

Gene Details

Gene
ARHGEF39
Gene Name
Rho guanine nucleotide exchange factor 39
There are no transcripts associated with this gene.

Gene
CA9
Gene Name
carbonic anhydrase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001216.2 277 Missense Mutation AGA,GGA R79G NP_001207.2
Gene
TPM2
Gene Name
tropomyosin 2 (beta)
There are no transcripts associated with this gene.

View Full Product Details