Product Details

SNP ID
rs114678429
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:6414847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTCTCTCTTTAAAATAATGTT[A/G]TAGCACCTTTGATTTGTGTTTGCAG
Phenotype
MIM: 615211
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UHRF2 PubMed Links

Gene Details

Gene
UHRF2
Gene Name
ubiquitin like with PHD and ring finger domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152896.2 Intron NP_690856.1
XM_011517704.2 Intron XP_011516006.1
XM_011517705.2 Intron XP_011516007.1
XM_017014253.1 Intron XP_016869742.1
XM_017014254.1 Intron XP_016869743.1

View Full Product Details