Product Details

SNP ID
rs116887436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:6533421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATAGCAAAGTTGTGTTTGAAAG[A/C]CTTCAACTGCTTAAAGAAGACTTTG
Phenotype
MIM: 238300
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GLDC PubMed Links

Gene Details

Gene
GLDC
Gene Name
glycine decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000170.2 Intron NP_000161.2

View Full Product Details