Product Details
- SNP ID
-
rs111524366
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:81585545 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGGGACAGCACGCAGCTCTCATG[C/T]AGGTGCAGCTGGTACTTGTCAGGCT
- Phenotype
-
MIM: 600189
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TLE1
PubMed Links
Gene Details
- Gene
- TLE1
- Gene Name
- transducin like enhancer of split 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001303103.1 |
2324 |
Silent Mutation |
CTA,CTG |
L706L |
NP_001290032.1 |
NM_001303104.1 |
2324 |
Silent Mutation |
CTA,CTG |
L681L |
NP_001290033.1 |
NM_005077.4 |
2324 |
Silent Mutation |
CTA,CTG |
L696L |
NP_005068.2 |
XM_005252151.1 |
2324 |
Silent Mutation |
CTA,CTG |
L740L |
XP_005252208.1 |
XM_005252152.1 |
2324 |
Silent Mutation |
CTA,CTG |
L739L |
XP_005252209.1 |
XM_005252153.1 |
2324 |
Silent Mutation |
CTA,CTG |
L730L |
XP_005252210.1 |
XM_005252154.1 |
2324 |
Silent Mutation |
CTA,CTG |
L729L |
XP_005252211.1 |
XM_005252156.2 |
2324 |
Silent Mutation |
CTA,CTG |
L705L |
XP_005252213.1 |
XM_005252162.1 |
2324 |
Silent Mutation |
CTA,CTG |
L656L |
XP_005252219.1 |
XM_005252163.2 |
2324 |
Silent Mutation |
CTA,CTG |
L622L |
XP_005252220.1 |
XM_006717258.1 |
2324 |
Silent Mutation |
CTA,CTG |
L739L |
XP_006717321.1 |
XM_006717259.3 |
2324 |
Silent Mutation |
CTA,CTG |
L684L |
XP_006717322.1 |
XM_006717260.1 |
2324 |
Silent Mutation |
CTA,CTG |
L666L |
XP_006717323.1 |
XM_006717261.2 |
2324 |
Silent Mutation |
CTA,CTG |
L625L |
XP_006717324.1 |
XM_006717262.1 |
2324 |
Silent Mutation |
CTA,CTG |
L695L |
XP_006717325.1 |
XM_006717263.1 |
2324 |
Silent Mutation |
CTA,CTG |
L694L |
XP_006717326.1 |
XM_011518951.2 |
2324 |
Silent Mutation |
CTA,CTG |
L695L |
XP_011517253.1 |
XM_017015064.1 |
2324 |
Silent Mutation |
CTA,CTG |
L705L |
XP_016870553.1 |
XM_017015065.1 |
2324 |
Silent Mutation |
CTA,CTG |
L704L |
XP_016870554.1 |
XM_017015066.1 |
2324 |
Silent Mutation |
CTA,CTG |
L650L |
XP_016870555.1 |
View Full Product Details