Product Details

SNP ID
rs115116010
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:90610067 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTATACACACTGTCCTTGTGGGGT[A/G]TGAATTCCAGGATGTGTCTTCAAGG
Phenotype
MIM: 607863
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIRAS2 PubMed Links

Gene Details

Gene
DIRAS2
Gene Name
DIRAS family GTPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017594.4 4151 UTR 3 NP_060064.2

View Full Product Details