Product Details

SNP ID
rs116489782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:96755915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTACATGGTACTGTAAAAGTACC[A/G]TGTACTTTTACCATGTCACCCAGTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF510 PubMed Links

Gene Details

Gene
ZNF510
Gene Name
zinc finger protein 510
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001314059.1 5187 UTR 3 NP_001300988.1
NM_001314060.1 5187 UTR 3 NP_001300989.1
NM_014930.2 5187 UTR 3 NP_055745.1
XM_005251808.2 5187 UTR 3 XP_005251865.1
XM_011518393.2 5187 UTR 3 XP_011516695.1
XM_017014483.1 5187 UTR 3 XP_016869972.1

View Full Product Details