Product Details

SNP ID
rs114599067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:99224282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATCCTGAATGATTTTGAGTTCC[C/T]TGTCATTGGGATGTTCATGAGGAGT
Phenotype
MIM: 607905 MIM: 609214
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALG2 PubMed Links

Gene Details

Gene
ALG2
Gene Name
ALG2, alpha-1,3/1,6-mannosyltransferase
There are no transcripts associated with this gene.

Gene
SEC61B
Gene Name
Sec61 translocon beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006808.2 Intron NP_006799.1

View Full Product Details