Product Details

SNP ID
rs113123140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:72056116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTACAGTACTGACCAGGTTCTG[C/T]CTGGCAGGTTCCCAGGTCTAAAAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf57 PubMed Links

Gene Details

Gene
C9orf57
Gene Name
chromosome 9 open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128618.1 244 Missense Mutation ACA,GCA T102A NP_001122090.1
XM_011518225.2 244 Missense Mutation ACA,GCA T52A XP_011516527.1

View Full Product Details