Product Details

SNP ID
rs118017926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:106864531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAGCTGTCTGCTTTGGTTCCAG[C/T]CCAGACTCCAAAACACAAAGCTCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105376204 PubMed Links

Gene Details

Gene
LOC105376204
Gene Name
uncharacterized LOC105376204
There are no transcripts associated with this gene.

Gene
ZNF462
Gene Name
zinc finger protein 462
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021224.4 385 Intron NP_067047.4
XM_006717209.3 385 Intron XP_006717272.1
XM_006717211.3 385 Intron XP_006717274.1
XM_006717212.3 385 Intron XP_006717275.1
XM_006717215.3 385 UTR 5 XP_006717278.1
XM_006717216.3 385 UTR 5 XP_006717279.1
XM_006717218.3 385 UTR 5 XP_006717281.1
XM_011518892.2 385 UTR 5 XP_011517194.1
XM_017014996.1 385 Intron XP_016870485.1
XM_017014997.1 385 Intron XP_016870486.1
XM_017014998.1 385 UTR 5 XP_016870487.1
XM_017014999.1 385 Intron XP_016870488.1

View Full Product Details