Product Details

SNP ID
rs116916279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114604073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTAAGGAACCAGCCCAAGGATG[G/T]GCAGTGATGGAGCCAAGATTCAAAC
Phenotype
MIM: 607296
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATP6V1G1 PubMed Links

Gene Details

Gene
ATP6V1G1
Gene Name
ATPase H+ transporting V1 subunit G1
There are no transcripts associated with this gene.

Gene
TMEM268
Gene Name
transmembrane protein 268
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153045.3 177 Intron NP_694590.2
XM_006716997.1 177 Intron XP_006717060.1
XM_011518350.1 177 Intron XP_011516652.1
XM_011518351.2 177 UTR 5 XP_011516653.1
XM_011518352.1 177 Intron XP_011516654.1
XM_011518353.1 177 Intron XP_011516655.1
XM_011518354.1 177 Intron XP_011516656.1
XM_011518355.1 177 Intron XP_011516657.1
XM_017014429.1 177 Intron XP_016869918.1
XM_017014430.1 177 Intron XP_016869919.1

View Full Product Details