Product Details

SNP ID
rs115294789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:72901123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTAAAAAAATCAAGAAAAGAAA[A/C]ATTTTGTCTTTAAAATCTACTATAT
Phenotype
MIM: 100640
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ALDH1A1 PubMed Links

Gene Details

Gene
ALDH1A1
Gene Name
aldehyde dehydrogenase 1 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000689.4 1908 UTR 3 NP_000680.2

View Full Product Details