Product Details

SNP ID
rs116547108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:68707749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCATCATTTCAGTTCTGATAGCA[A/G]TGAGGACACATCATGGAAACTGCAA
Phenotype
MIM: 602745
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIP5K1B PubMed Links

Gene Details

Gene
PIP5K1B
Gene Name
phosphatidylinositol-4-phosphate 5-kinase type 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278253.1 Intron NP_001265182.1
NM_003558.3 Intron NP_003549.1
XM_005252261.3 Intron XP_005252318.1
XM_005252262.4 Intron XP_005252319.1
XM_006717300.2 Intron XP_006717363.1
XM_006717301.1 Intron XP_006717364.1
XM_011519082.2 Intron XP_011517384.1
XM_011519083.2 Intron XP_011517385.1
XM_011519084.2 Intron XP_011517386.1
XM_017015188.1 Intron XP_016870677.1
XM_017015189.1 Intron XP_016870678.1
XM_017015190.1 Intron XP_016870679.1
XM_017015191.1 Intron XP_016870680.1

View Full Product Details