Product Details

SNP ID
rs112623977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:125153582 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCAACCTGCTCCTCGGGGACTG[A/G]TAGCCCAGGTATCCACATCTTCAGG
Phenotype
MIM: 612725
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPP6C PubMed Links

Gene Details

Gene
PPP6C
Gene Name
protein phosphatase 6 catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123355.1 638 Missense Mutation ACC,ATC T244I NP_001116827.1
NM_001123369.1 638 Missense Mutation ACC,ATC T185I NP_001116841.1
NM_002721.4 638 Missense Mutation ACC,ATC T207I NP_002712.1
XM_011518847.2 638 Missense Mutation ACC,ATC T195I XP_011517149.1

View Full Product Details