Product Details

SNP ID
rs117863659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:125285227 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGCTTCATTCCAATACAACTTT[C/T]ATTTATACAAACAGGCAGCAGGCTA
Phenotype
MIM: 611714
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAPVD1 PubMed Links

Gene Details

Gene
GAPVD1
Gene Name
GTPase activating protein and VPS9 domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282679.1 Intron NP_001269608.1
NM_001282680.1 Intron NP_001269609.1
NM_001282681.1 Intron NP_001269610.1
NM_015635.3 Intron NP_056450.2
XM_005251901.3 Intron XP_005251958.1
XM_005251904.3 Intron XP_005251961.1
XM_006717044.3 Intron XP_006717107.1
XM_011518499.2 Intron XP_011516801.1
XM_011518500.2 Intron XP_011516802.1
XM_011518502.2 Intron XP_011516804.1
XM_011518506.2 Intron XP_011516808.1
XM_011518507.2 Intron XP_011516809.1
XM_017014600.1 Intron XP_016870089.1
XM_017014601.1 Intron XP_016870090.1
XM_017014602.1 Intron XP_016870091.1
XM_017014603.1 Intron XP_016870092.1
XM_017014604.1 Intron XP_016870093.1
XM_017014605.1 Intron XP_016870094.1
XM_017014606.1 Intron XP_016870095.1
XM_017014607.1 Intron XP_016870096.1
XM_017014608.1 Intron XP_016870097.1
XM_017014609.1 Intron XP_016870098.1
XM_017014610.1 Intron XP_016870099.1
XM_017014611.1 Intron XP_016870100.1

View Full Product Details