Product Details

SNP ID
rs111622564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128422925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGATGCTGCAGGAGTGGCTGGC[A/G]GCTGTGGGCGATGACTATGCTGCTG
Phenotype
MIM: 616626
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CERCAM PubMed Links

Gene Details

Gene
CERCAM
Gene Name
cerebral endothelial cell adhesion molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286760.1 653 Silent Mutation GCA,GCG A7A NP_001273689.1
NM_016174.4 653 Silent Mutation GCA,GCG A85A NP_057258.3
XM_005252035.1 653 Silent Mutation GCA,GCG A85A XP_005252092.1
XM_011518762.1 653 Silent Mutation GCA,GCG A85A XP_011517064.1
XM_011518763.2 653 Silent Mutation GCA,GCG A7A XP_011517065.1
XM_017014794.1 653 Silent Mutation GCA,GCG A85A XP_016870283.1

View Full Product Details