Product Details

SNP ID
rs111700515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128731396 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTGCAGTGCTCAATCACCTTGC[G/T]GGAGACAATGGGGGAGACAGGATTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100506100 PubMed Links

Gene Details

Gene
LOC100506100
Gene Name
uncharacterized LOC100506100
There are no transcripts associated with this gene.

Gene
ZDHHC12
Gene Name
zinc finger DHHC-type containing 12
There are no transcripts associated with this gene.

Gene
ZER1
Gene Name
zyg-11 related cell cycle regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006336.3 Intron NP_006327.2
XM_005251645.2 Intron XP_005251702.1
XM_005251646.2 Intron XP_005251703.1
XM_011518122.2 Intron XP_011516424.1
XM_017014185.1 Intron XP_016869674.1
XM_017014186.1 Intron XP_016869675.1
XM_017014187.1 Intron XP_016869676.1
XM_017014188.1 Intron XP_016869677.1
XM_017014189.1 Intron XP_016869678.1
XM_017014190.1 Intron XP_016869679.1

View Full Product Details