Product Details

SNP ID
rs118092994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:130054511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAGGTACTTGGAACCTTAGCCT[C/T]AGTTTCCCATTTCGGTTTAGGGCGC
Phenotype
MIM: 606191
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FNBP1 PubMed Links

Gene Details

Gene
FNBP1
Gene Name
formin binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015033.2 Intron NP_055848.1
XM_005251815.2 Intron XP_005251872.1
XM_005251820.2 Intron XP_005251877.1
XM_005251821.2 Intron XP_005251878.1
XM_005251822.2 Intron XP_005251879.1
XM_005251823.2 Intron XP_005251880.1
XM_005251824.2 Intron XP_005251881.1
XM_005251825.2 Intron XP_005251882.1
XM_005251826.2 Intron XP_005251883.1
XM_005251827.2 Intron XP_005251884.1
XM_005251828.2 Intron XP_005251885.1
XM_005251829.2 Intron XP_005251886.1
XM_005251830.2 Intron XP_005251887.1
XM_005251831.2 Intron XP_005251888.1
XM_005251832.2 Intron XP_005251889.1
XM_005251833.2 Intron XP_005251890.1
XM_005251834.2 Intron XP_005251891.1
XM_006717016.2 Intron XP_006717079.1
XM_011518399.2 Intron XP_011516701.1
XM_011518400.2 Intron XP_011516702.1
XM_011518401.1 Intron XP_011516703.1
XM_011518402.1 Intron XP_011516704.1
XM_017014487.1 Intron XP_016869976.1
XM_017014488.1 Intron XP_016869977.1
XM_017014489.1 Intron XP_016869978.1
XM_017014490.1 Intron XP_016869979.1
XM_017014491.1 Intron XP_016869980.1
XM_017014492.1 Intron XP_016869981.1
XM_017014493.1 Intron XP_016869982.1
XM_017014494.1 Intron XP_016869983.1
Gene
GPR107
Gene Name
G protein-coupled receptor 107
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136557.1 Intron NP_001130029.1
NM_001136558.1 Intron NP_001130030.1
NM_001287346.1 Intron NP_001274275.1
NM_020960.4 Intron NP_066011.2
XM_017014953.1 Intron XP_016870442.1
XM_017014954.1 Intron XP_016870443.1

View Full Product Details