Product Details

SNP ID
rs116831514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:76612317 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTAAAAATATATATATAAATGGT[A/G]AGATTGTTGCTGTGTCTGTTTGGCT
Phenotype
MIM: 610691
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRUNE2 PubMed Links

Gene Details

Gene
PRUNE2
Gene Name
prune homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308047.1 11786 UTR 3 NP_001294976.1
NM_001308048.1 11786 UTR 3 NP_001294977.1
NM_001308049.1 11786 UTR 3 NP_001294978.1
NM_001308050.1 11786 UTR 3 NP_001294979.1
NM_001308051.1 11786 UTR 3 NP_001294980.1
NM_015225.2 11786 UTR 3 NP_056040.2
XM_005251745.1 11786 UTR 3 XP_005251802.1
XM_005251746.1 11786 UTR 3 XP_005251803.1
XM_005251748.1 11786 UTR 3 XP_005251805.1
XM_005251750.1 11786 UTR 3 XP_005251807.1
XM_005251751.1 11786 UTR 3 XP_005251808.1
XM_005251754.1 11786 UTR 3 XP_005251811.1
XM_006716982.1 11786 UTR 3 XP_006717045.1
XM_006716983.1 11786 UTR 3 XP_006717046.1
XM_006716984.2 11786 Intron XP_006717047.1
XM_006716985.1 11786 UTR 3 XP_006717048.1
XM_006716986.1 11786 UTR 3 XP_006717049.1
XM_011518323.1 11786 UTR 3 XP_011516625.1
XM_011518326.2 11786 Intron XP_011516628.1
XM_011518327.1 11786 UTR 3 XP_011516629.1
XM_011518328.2 11786 Intron XP_011516630.1
XM_017014345.1 11786 UTR 3 XP_016869834.1
XM_017014346.1 11786 UTR 3 XP_016869835.1
XM_017014347.1 11786 UTR 3 XP_016869836.1
XM_017014348.1 11786 UTR 3 XP_016869837.1
XM_017014349.1 11786 UTR 3 XP_016869838.1
XM_017014350.1 11786 UTR 3 XP_016869839.1
XM_017014351.1 11786 UTR 3 XP_016869840.1
XM_017014352.1 11786 UTR 3 XP_016869841.1
XM_017014353.1 11786 Intron XP_016869842.1
XM_017014354.1 11786 UTR 3 XP_016869843.1
XM_017014355.1 11786 UTR 3 XP_016869844.1
XM_017014356.1 11786 UTR 3 XP_016869845.1
XM_017014357.1 11786 UTR 3 XP_016869846.1
XM_017014358.1 11786 UTR 3 XP_016869847.1
XM_017014359.1 11786 UTR 3 XP_016869848.1

View Full Product Details