Product Details

SNP ID
rs112264167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:10558993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGTCCTGAGCTGATTCCAGTTA[C/G]GAACCAAGGGTGGACTTTCTTCTTT
Phenotype
MIM: 605702
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LYVE1 PubMed Links

Gene Details

Gene
LYVE1
Gene Name
lymphatic vessel endothelial hyaluronan receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006691.3 1373 UTR 3 NP_006682.2
Gene
MRVI1-AS1
Gene Name
MRVI1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details