Product Details

SNP ID
rs115342209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:15120526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGAAGAGGAGGACATTGTAGGT[A/G]GAGAGACCAGCACGTGTGAGGGCAC
Phenotype
MIM: 610668
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
INSC PubMed Links

Gene Details

Gene
INSC
Gene Name
inscuteable homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031853.4 Intron NP_001027024.3
NM_001042536.2 Intron NP_001036001.1
NM_001278313.1 Intron NP_001265242.1
NM_001278314.1 Intron NP_001265243.1
NM_001278315.1 Intron NP_001265244.1
NM_001278316.1 Intron NP_001265245.1
XM_006718227.3 Intron XP_006718290.1
XM_011520085.1 Intron XP_011518387.1
XM_011520086.1 Intron XP_011518388.1
XM_017017697.1 Intron XP_016873186.1
XM_017017698.1 Intron XP_016873187.1

View Full Product Details