Product Details
- SNP ID
-
rs117553480
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:15969492 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTGTGGACAGTGGGTTCTGAACT[G/T]CATGAGAACCCTGGTGAAACGTTTT
- Phenotype
-
MIM: 607257
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MIR6073
PubMed Links
Gene Details
- Gene
- MIR6073
- Gene Name
- microRNA 6073
There are no transcripts associated with this gene.
- Gene
- SOX6
- Gene Name
- SRY-box 6
View Full Product Details