Product Details
- SNP ID
-
rs115211278
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:17555538 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGGAATCTGAGCCATCCAGACC[A/G]GGGCTAGAATCCCAGCTCTGGGATT
- Phenotype
-
MIM: 604487
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OTOG
PubMed Links
Gene Details
- Gene
- OTOG
- Gene Name
- otogelin
View Full Product Details