Product Details

SNP ID
rs117297841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18284381 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATCAGCATGTCCCACTGCATTT[G/T]CCTTTGAAATGCCCACTGGATGTGC
Phenotype
MIM: 607521
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HPS5 PubMed Links

Gene Details

Gene
HPS5
Gene Name
HPS5, biogenesis of lysosomal organelles complex 2 subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007216.3 Intron NP_009147.3
NM_181507.1 Intron NP_852608.1
NM_181508.1 Intron NP_852609.1
XM_011519862.1 Intron XP_011518164.1
XM_011519863.1 Intron XP_011518165.1
XM_011519864.1 Intron XP_011518166.1
XM_011519865.1 Intron XP_011518167.1
XM_011519866.1 Intron XP_011518168.1
XM_011519867.1 Intron XP_011518169.1
XM_011519868.2 Intron XP_011518170.1
XM_017017149.1 Intron XP_016872638.1
XM_017017150.1 Intron XP_016872639.1
XM_017017151.1 Intron XP_016872640.1
XM_017017152.1 Intron XP_016872641.1
XM_017017153.1 Intron XP_016872642.1
XM_017017154.1 Intron XP_016872643.1

View Full Product Details