Product Details
- SNP ID
-
rs117533054
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:19350821 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAAGACAGGAAGACCTTTGAAGA[C/G]GTGGTGCTGATTTCCTTGGCTGGCG
- Phenotype
-
MIM: 607026
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NAV2
PubMed Links
Gene Details
- Gene
- NAV2
- Gene Name
- neuron navigator 2
View Full Product Details