Product Details

SNP ID
rs115266356
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:373061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTGAAGAAGTTGGCCGTGTCCCC[C/T]AAGTGGAAGAACTATGGACTCCGTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B4GALNT4 PubMed Links

Gene Details

Gene
B4GALNT4
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178537.4 489 Silent Mutation CCC,CCT P160P NP_848632.2
XM_017017654.1 489 Silent Mutation CCC,CCT P68P XP_016873143.1

View Full Product Details