Product Details

SNP ID
rs117698578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3856526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAGGTTCATGGAGGATTCACAC[A/C]TGGCACTGCCTGTTCCAAGTAGTTC
Phenotype
MIM: 605921
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MIR4687 PubMed Links

Gene Details

Gene
MIR4687
Gene Name
microRNA 4687
There are no transcripts associated with this gene.

Gene
STIM1
Gene Name
stromal interaction molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277961.1 Intron NP_001264890.1
NM_001277962.1 Intron NP_001264891.1
NM_003156.3 Intron NP_003147.2

View Full Product Details