Product Details

SNP ID
rs115370823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:4595000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCACTTCCTCTTTGACCACTCCA[A/G]CCTGGGTTCATGAACACAATATCTG
Phenotype
MIM: 613184
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52I1 PubMed Links

Gene Details

Gene
OR52I1
Gene Name
olfactory receptor family 52 subfamily I member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005169.1 962 Missense Mutation AAC,AGC N321S NP_001005169.1
Gene
OR52I2
Gene Name
olfactory receptor family 52 subfamily I member 2
There are no transcripts associated with this gene.

Gene
TRIM68
Gene Name
tripartite motif containing 68
There are no transcripts associated with this gene.

View Full Product Details