Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_198075.3 | 120 | Intron | NP_932341.1 | ||
XM_005252775.2 | 120 | Intron | XP_005252832.1 | ||
XM_005252776.2 | 120 | Intron | XP_005252833.1 | ||
XM_006718132.2 | 120 | Intron | XP_006718195.1 | ||
XM_006718133.2 | 120 | Intron | XP_006718196.1 | ||
XM_011519875.2 | 120 | Intron | XP_011518177.1 | ||
XM_011519876.1 | 120 | Intron | XP_011518178.1 | ||
XM_011519877.2 | 120 | Intron | XP_011518179.1 | ||
XM_017017167.1 | 120 | Intron | XP_016872656.1 | ||
XM_017017168.1 | 120 | UTR 5 | XP_016872657.1 |