Product Details
- SNP ID
-
rs116121413
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:47274631 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCGATACCCCTTGGAGGATCACA[A/C]TGAGTTTCCCCTGCCCCCAGATGTA
- Phenotype
-
MIM: 603584
MIM: 602423
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LOC101928943
PubMed Links
Gene Details
- Gene
- LOC101928943
- Gene Name
- uncharacterized LOC101928943
There are no transcripts associated with this gene.
- Gene
- MADD
- Gene Name
- MAP kinase activating death domain
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001135943.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
NP_001129415.1 |
NM_001135944.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
NP_001129416.1 |
NM_003682.3 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
NP_003673.3 |
NM_130470.2 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
NP_569826.2 |
NM_130471.2 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
NP_569827.2 |
NM_130472.2 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
NP_569828.2 |
NM_130473.2 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
NP_569829.2 |
NM_130474.2 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
NP_569830.2 |
NM_130475.2 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
NP_569831.1 |
NM_130476.2 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
NP_569832.2 |
XM_005253189.2 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_005253246.1 |
XM_005253196.2 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_005253253.1 |
XM_005253199.2 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_005253256.1 |
XM_005253200.2 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_005253257.1 |
XM_005253201.2 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_005253258.1 |
XM_005253203.2 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_005253260.1 |
XM_005253204.2 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_005253261.1 |
XM_005253205.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_005253262.1 |
XM_017018478.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873967.1 |
XM_017018479.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873968.1 |
XM_017018480.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873969.1 |
XM_017018481.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873970.1 |
XM_017018482.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873971.1 |
XM_017018483.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873972.1 |
XM_017018484.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873973.1 |
XM_017018485.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873974.1 |
XM_017018486.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873975.1 |
XM_017018487.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873976.1 |
XM_017018488.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873977.1 |
XM_017018489.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873978.1 |
XM_017018490.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873979.1 |
XM_017018491.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873980.1 |
XM_017018492.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873981.1 |
XM_017018493.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873982.1 |
XM_017018494.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873983.1 |
XM_017018495.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873984.1 |
XM_017018496.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873985.1 |
XM_017018497.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873986.1 |
XM_017018498.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873987.1 |
XM_017018499.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873988.1 |
XM_017018500.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873989.1 |
XM_017018501.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873990.1 |
XM_017018502.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873991.1 |
XM_017018503.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873992.1 |
XM_017018504.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873993.1 |
XM_017018505.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873994.1 |
XM_017018506.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873995.1 |
XM_017018507.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873996.1 |
XM_017018508.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873997.1 |
XM_017018509.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873998.1 |
XM_017018510.1 |
346 |
Missense Mutation |
AAT,ACT |
N44T |
XP_016873999.1 |
- Gene
- NR1H3
- Gene Name
- nuclear receptor subfamily 1 group H member 3
There are no transcripts associated with this gene.
View Full Product Details