Product Details

SNP ID
rs112604883
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5131761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTTGCTTGGTCTTCACTCCATA[A/G]ACAATAGGGTTGAGAAAAGGTGGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52A5 PubMed Links

Gene Details

Gene
OR52A5
Gene Name
olfactory receptor family 52 subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005160.2 882 Silent Mutation GTC,GTT V294V NP_001005160.1

View Full Product Details