Product Details

SNP ID
rs117284118
Assay Type
Functionally Tested
NCBI dbSNP Submissions
16
Location
Chr.11:5351894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTACCATCCGCAGCCCCTTAAG[A/C]TATACCTCTATCCTGACCAACACCC
Phenotype
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005567.2 387 Intron NP_001005567.2
Gene
OR51B6
Gene Name
olfactory receptor family 51 subfamily B member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004750.1 387 Missense Mutation AGA,AGC R129S NP_001004750.1

View Full Product Details