Product Details

SNP ID
rs113436707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5422249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGAAGGCATCTACTTCATCCTC[A/G]CGGACATCCCTGGATTTGAGGCCTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR51B5 PubMed Links

Gene Details

Gene
OR51B5
Gene Name
olfactory receptor family 51 subfamily B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005567.2 139 Intron NP_001005567.2
Gene
OR51Q1
Gene Name
olfactory receptor family 51 subfamily Q member 1 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004757.2 139 Missense Mutation ACG,GCG T17A NP_001004757.1

View Full Product Details