Product Details
- SNP ID
-
rs117935003
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:5515285 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTTGAAGCTGCTGGGTAAGCTCT[A/G]TGCTAGGTAAGGCTGGTATCCAAGC
- Phenotype
-
MIM: 605473
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OLFM5P
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs393044] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OLFM5P
- Gene Name
- olfactomedin family member 5, pseudogene
There are no transcripts associated with this gene.
- Gene
- OR51B5
- Gene Name
- olfactory receptor family 51 subfamily B member 5
There are no transcripts associated with this gene.
- Gene
- UBQLN3
- Gene Name
- ubiquilin 3
There are no transcripts associated with this gene.
- Gene
- UBQLNL
- Gene Name
- ubiquilin-like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145053.4 |
1442 |
Missense Mutation |
ACA,ATA |
T386I |
NP_659490.4 |
View Full Product Details