Product Details

SNP ID
rs114930867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5544695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAACATGATGTGGAAGGTGCGAG[A/G]GACATTGTGTCCAAAGCGATGGGCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR52H1 PubMed Links

Gene Details

Gene
OR52H1
Gene Name
olfactory receptor family 52 subfamily H member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005289.1 829 Missense Mutation CCT,TCT P277S NP_001005289.1

View Full Product Details