Product Details

SNP ID
rs116991228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:5724007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACTCTCCCTGCATAATGTGGAAT[C/T]CAGATCATGGCAGGAGCTACTTTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR52N4 PubMed Links

Gene Details

Gene
OR52N4
Gene Name
olfactory receptor family 52 subfamily N member 4 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005175.3 1871 Intron NP_001005175.3
XM_017017711.1 1871 UTR 5 XP_016873200.1
XM_017017712.1 1871 UTR 5 XP_016873201.1
XM_017017713.1 1871 UTR 5 XP_016873202.1

View Full Product Details