Product Details

SNP ID
rs114783985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6846371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGCTGCTGCTATCCTCAAGATCC[C/T]ATCAGCTAAAGGGAAGCATAAAGCC
Phenotype
MIM: 608493
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10A5 PubMed Links

Gene Details

Gene
OR10A5
Gene Name
olfactory receptor family 10 subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178168.1 689 Missense Mutation CCA,CTA P230L NP_835462.1

View Full Product Details