Product Details
- SNP ID
-
rs112948624
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:748344 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGCCGGTGCCCATGAAAGCACAG[A/G]GTTGGTACACTGTTAAATACTGGAT
- Phenotype
-
MIM: 602063
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TALDO1
PubMed Links
Gene Details
- Gene
- TALDO1
- Gene Name
- transaldolase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006755.1 |
|
Intron |
|
|
NP_006746.1 |
View Full Product Details