Product Details

SNP ID
rs113981530
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:9023115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTCTAGCTTACTTCCTATAGGCA[C/T]CATTCAAATTAAGCATATGCAAAGC
Phenotype
MIM: 611747
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCUBE2 PubMed Links

Gene Details

Gene
SCUBE2
Gene Name
signal peptide, CUB domain and EGF like domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170690.1 3897 Intron NP_001164161.1
NM_020974.2 3897 Intron NP_066025.2
XM_005253032.4 3897 Intron XP_005253089.2
XM_005253033.4 3897 Intron XP_005253090.2
XM_005253034.4 3897 Intron XP_005253091.2
XM_005253035.4 3897 UTR 3 XP_005253092.2
XM_005253036.4 3897 Intron XP_005253093.2
XM_005253037.4 3897 Intron XP_005253094.2
XM_011520246.2 3897 Intron XP_011518548.1
XM_011520247.2 3897 Intron XP_011518549.1
XM_011520248.2 3897 Intron XP_011518550.1
XM_017018080.1 3897 Intron XP_016873569.1
XM_017018081.1 3897 Intron XP_016873570.1
XM_017018082.1 3897 UTR 3 XP_016873571.1

View Full Product Details