Product Details

SNP ID
rs104894360
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:25209904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGAACTAATGTATAGAAGGCATCA[T/A]CAACACCCTGAAATACATAAAAAGT
Phenotype
MIM: 190070
Polymorphism
T/A, Transversion substitution
Allele Nomenclature
Literature Links
KRAS PubMed Links

Gene Details

Gene
KRAS
Gene Name
KRAS proto-oncogene, GTPase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004985.4 780 Missense Mutation GAT,GTT D153V NP_004976.2
NM_033360.3 780 UTR 3 NP_203524.1
XM_006719069.3 780 UTR 3 XP_006719132.1
XM_011520653.2 780 Intron XP_011518955.1
XM_017019292.1 780 Intron XP_016874781.1
XM_017019293.1 780 Missense Mutation GAT,GTT D192V XP_016874782.1
Gene
LYRM5
Gene Name
LYR motif containing 5
There are no transcripts associated with this gene.

View Full Product Details