Product Details

SNP ID
rs112247122
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:29153423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTAAGAGAAAACGCTTGCAGCA[C/T]TCAGGGGACAGAGAGAAGGCAGGCC
Phenotype
MIM: 616156
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAR2 PubMed Links

Gene Details

Gene
FAR2
Gene Name
fatty acyl-CoA reductase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271783.1 Intron NP_001258712.1
NM_001271784.1 Intron NP_001258713.1
NM_018099.4 Intron NP_060569.3
XM_011520747.2 Intron XP_011519049.1
XM_011520748.2 Intron XP_011519050.1
XM_017019624.1 Intron XP_016875113.1
XM_017019625.1 Intron XP_016875114.1

View Full Product Details